siRNA Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147201.2 | NP_671734.1 | 4 | 456 | ||
| GenBank mRNA | AF302138.1 | - | 4 | 455 | ||
| AK046142.1 | - | 4 | 450 | |||
| AK077440.1 | - | 4 | 492 | |||
| AK170501.1 | - | 4 | 472 | |||
| AK178908.1 | - | 4 | 297 | |||
| AK191367.1 | - | 5 | 335 | |||
| BC004756.1 | - | 4 | 426 | 3154089 | ||
| BC018463.1 | - | 4 | 430 | 3598438 | ||

Nrbp1
Genomic Map