siRNA Details
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147201.2 | NP_671734.1 | 12 | 1188 | ||
GenBank mRNA | AF302138.1 | - | 12 | 1187 | ||
AK046142.1 | - | 13 | 1206 | |||
AK077440.1 | - | 12 | 1224 | |||
AK170501.1 | - | 12 | 1204 | |||
AK207083.1 | - | 4 | 329 | |||
AK215481.1 | - | 1 | 34 | |||
BC004756.1 | - | 12 | 1158 | 3154089 | ||
BC013212.1 | - | 1 | 582 | |||
BC018463.1 | - | 12 | 1162 | 3598438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK041029.1 | - | 1253 | Chr. - - [-] on Build |
