siRNA Details
| Gene Symbol: Zfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177565.3 | NP_808233.1 | 2 | 1056 | ||
| GenBank mRNA | AK045753.1 | - | 2 | 1057 | ||
| BC032259.1 | - | 1 | 225 | |||
| BC096463.1 | - | 2 | 1038 | 6466366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp3| Gene Symbol: Zfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177565.3 | NP_808233.1 | 2 | 1056 | ||
| GenBank mRNA | AK045753.1 | - | 2 | 1057 | ||
| BC032259.1 | - | 1 | 225 | |||
| BC096463.1 | - | 2 | 1038 | 6466366 | ||