siRNA Details
| Gene Symbol: Rac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009008.3 | NP_033034.1 | 3, 4 | 363 | ||
| GenBank mRNA | AK007561.1 | - | 3, 4 | 301 | ||
| AK011045.1 | - | 3, 4 | 342 | |||
| AK088415.1 | - | 3, 4 | 312 | |||
| AK088805.1 | - | 3, 4 | 300 | |||
| AK144136.1 | - | 3, 4 | 307 | |||
| AK171321.1 | - | 3, 4 | 364 | |||
| BC005455.1 | - | 3, 4 | 279 | 3495818 | ||
| X53247.1 | - | 3, 4 | 261 | |||

Rac2
Genomic Map