siRNA Details
| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001176528.1 | NP_001169999.1 | 6 | 1377 | ||
| NM_001177302.1 | NP_001170773.1 | 8 | 1582 | |||
| NM_001177303.1 | NP_001170774.1 | 7 | 1327 | |||
| NM_009024.2 | NP_033050.2 | 7 | 1449 | |||
| GenBank mRNA | AB221572.1 | - | 6 | 897 | ||
| AK042163.1 | - | 7 | 1328 | |||
| AK153644.1 | - | 6 | 923 | |||
| AK154623.1 | - | 7 | 1422 | |||
| AK170956.1 | - | 6 | 1377 | |||
| BC010216.1 | - | 7 | 1447 | 3498142 | ||
| BC038266.1 | - | 6 | 1446 | |||
| BC040383.1 | - | 8 | 1582 | |||
| CT010231.1 | - | 6 | 892 | |||
| S56656.1 | - | 7 | 1429 | |||

Rara
Genomic Map