siRNA Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007572.2 | NP_001007573.1 | 1 | 153 | ||
| GenBank mRNA | AK085479.1 | - | 1 | 199 | ||
| BC110554.1 | - | 1 | 153 | 40047811 | ||
| BC110555.1 | - | 1 | 153 | 40047816 | ||

Trpv5
Genomic Map