siRNA Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166623.1 | NP_001160095.1 | 2 | 290 | ||
| NM_011252.4 | NP_035382.1 | 2 | 287 | |||
| GenBank mRNA | AJ237846.1 | - | 2 | 217 | ||
| AJ237847.1 | - | 2 | 232 | |||
| AK083899.1 | - | 2 | 287 | |||
| AK135164.1 | - | 2 | 292 | |||
| AK165006.1 | - | 2 | 233 | |||
| AK207357.1 | - | 2 | 151 | |||
| BC003710.1 | - | 2 | 285 | 3599678 | ||
| BY279114.1 | - | 2 | 287 | |||
| CK619886.1 | - | 1 | 93 | |||

Rbmx
Genomic Map