siRNA Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009034.4 | NP_033060.3 | 2 | 194 | ||
| GenBank mRNA | AK003312.1 | - | 2 | 179 | ||
| BC106846.2 | - | 2 | 203 | |||
| BC125551.1 | - | 2 | 163 | 40130066 | ||
| BC125553.1 | - | 2 | 163 | 40130068 | ||
| X74154.1 | - | 2 | 178 | |||

Rbp2
Genomic Map