siRNA Details
| Gene Symbol: Rcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009037.2 | NP_033063.1 | 2 | 467 | ||
| GenBank mRNA | AK017494.1 | - | 2 | 374 | ||
| AK133971.1 | - | 2 | 417 | |||
| AK159973.1 | - | 2 | 370 | |||
| AK214949.1 | - | 1 | 99 | |||
| BC049108.1 | - | 2 | 360 | 6414128 | ||
| D13003.1 | - | 2 | 387 | |||

Rcn1
Genomic Map