siRNA Details
Gene Symbol: Reg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009043.1 | NP_033069.1 | 4 | 343 | ||
GenBank mRNA | AK008196.1 | - | 4 | 342 | ||
BC098503.1 | - | 3 | 326 | 6431377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Reg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009043.1 | NP_033069.1 | 4 | 343 | ||
GenBank mRNA | AK008196.1 | - | 4 | 342 | ||
BC098503.1 | - | 3 | 326 | 6431377 |