siRNA Details
| Gene Symbol: Slc50a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009057.3 | NP_033083.2 | 4 | 499 | ||
| GenBank mRNA | AK014305.1 | - | 4 | 452 | ||
| AK045924.1 | - | 2 | 812 | |||
| AK079716.1 | - | 3 | 564 | |||
| AK158862.1 | - | 3 | 524 | |||
| AK170162.1 | - | 4 | 450 | |||
| BC014292.1 | - | 4 | 483 | 4190124 | ||
| X96618.1 | - | 4 | 490 | |||

Slc50a1
Genomic Map