siRNA Details
| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029987.2 | NP_084263.2 | 10 | 1094 | ||
| GenBank mRNA | AF410461.1 | - | 9 | 1042 | ||
| BC090656.1 | - | 10 | 1070 | |||
| BC130028.1 | - | 9 | 1046 | 40140883 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpe65| Gene Symbol: Rpe65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029987.2 | NP_084263.2 | 10 | 1094 | ||
| GenBank mRNA | AF410461.1 | - | 9 | 1042 | ||
| BC090656.1 | - | 10 | 1070 | |||
| BC130028.1 | - | 9 | 1046 | 40140883 | ||