siRNA Details
| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009086.2 | NP_033112.2 | 5 | 793 | ||
| GenBank mRNA | AK052672.1 | - | 5 | 763 | ||
| AK083732.1 | - | 5 | 775 | |||
| AK143501.1 | - | 5 | 730 | |||
| AK144616.1 | - | 5 | 780 | |||
| AK156712.1 | - | 5 | 784 | |||
| BC060656.1 | - | 5 | 723 | 30355130 | ||
| U58280.1 | - | 5 | 757 | |||

Polr1b
Genomic Map