siRNA Details
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025364.3 | NP_001020535.1 | 6 | 612 | ||
NM_013648.6 | NP_038676.1 | 9 | 1526 | |||
GenBank mRNA | AF038538.1 | - | 9 | 1482 | ||
AF038539.1 | - | 6 | 551 | |||
AK144271.1 | - | 9 | 1489 | |||
AK160500.1 | - | 6 | 573 | |||
BC031370.1 | - | 9 | 1483 | 5359358 |
