siRNA Details
Gene Symbol: Sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014418.1 | - | 4 | 660 | ||
AK157860.1 | - | 4 | 525 | |||
AK163400.1 | - | 4 | 645 | |||
AK167113.1 | - | 4 | 574 | |||
BC005715.1 | - | 5 | 794 | |||
BC068152.1 | - | 4 | 900 | 30534982 | ||
D50461.1 | - | 4 | 903 | |||
U45977.1 | - | 4 | 808 | |||
U45978.1 | - | 4 | 676 |
