siRNA Details
| Gene Symbol: Sepw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009156.2 | NP_033182.1 | 1, 2 | 113 | ||
| GenBank mRNA | AF015284.1 | - | 1, 2 | 68 | ||
| AF241527.2 | - | 1, 2 | 66 | |||
| AK002870.1 | - | 1, 2 | 89 | |||
| AK028124.1 | - | 1, 2 | 83 | |||
| BC052719.1 | - | 1, 2 | 75 | 6814242 | ||
| BU604713.1 | - | 1, 2 | 146 | |||
| U67890.1 | - | 1, 2 | 65 | |||

Sepw1
Genomic Map