siRNA Details
| Gene Symbol: Shh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009170.3 | NP_033196.1 | 1 | 548 | ||
| GenBank mRNA | AK077688.1 | - | 1 | 549 | ||
| BC063087.1 | - | 1 | 535 | 6516263 | ||
| EU664592.1 | - | 1 | 94 | |||
| X76290.1 | - | 1 | 210 | |||
| Gene Symbol: 9530036O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035417.1 | - | 1 | 55 | ||
| AK162496.1 | - | 177 | ||||

Shh
Genomic Map