siRNA Details
| Gene Symbol: St6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252505.1 | NP_001239434.1 | 7, 8 | 1454 | ||
| NM_001252506.1 | NP_001239435.1 | 6, 7 | 1321 | |||
| NM_145933.4 | NP_666045.1 | 7, 8 | 1785 | |||
| GenBank mRNA | AK034768.1 | - | 1, 2 | 131 | ||
| AK084124.1 | - | 7, 8 | 1620 | |||
| BC027833.1 | - | 4, 5 | 982 | 5321763 | ||
| BC092222.1 | - | 7, 8 | 1763 | 6844069 | ||
| BC096026.1 | - | 7, 8 | 1433 | 6833540 | ||
| CF741963.1 | - | 4, 5 | 416 | |||
| D16106.1 | - | 6, 7 | 1272 | |||

St6gal1
Genomic Map