siRNA Details
| Gene Symbol: St3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009179.3 | NP_033205.2 | 3, 4 | 1721 | ||
| GenBank mRNA | AK029246.1 | - | 3, 4 | 1636 | ||
| AK031052.1 | - | 3, 4 | 1664 | |||
| AK034863.1 | - | 3, 4 | 1662 | |||
| AK053827.1 | - | 3, 4 | 1660 | |||
| AK131653.1 | - | 2, 3 | 544 | |||
| BC066064.1 | - | 3, 4 | 1620 | 6850583 | ||
| X76989.1 | - | 2, 3 | 627 | |||

St3gal2
Genomic Map