siRNA Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009203.3 | NP_033229.3 | 10 | 1955 | ||
| GenBank mRNA | AB005451.1 | - | 10 | 1736 | ||
| AI314749.1 | - | 1 | 272 | |||
| AK142580.1 | - | 9 | 1104 | |||
| BC035927.1 | - | 1891 | 4242354 | |||

Slc22a12
Genomic Map