siRNA Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009204.2 | NP_033230.2 | 3 | 517 | ||
| GenBank mRNA | AB008453.1 | - | 3 | 315 | ||
| AK137607.1 | - | 3 | 502 | |||
| AK138832.1 | - | 3 | 491 | |||
| AK160882.1 | - | 3 | 503 | |||
| AK164310.1 | - | 3 | 492 | |||
| BC014282.1 | - | 3 | 487 | 4207674 | ||
| M23383.1 | - | 3 | 467 | |||

Slc2a4
Genomic Map