siRNA Details
| Gene Symbol: Slfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011409.1 | NP_035539.1 | 2 | 547 | ||
| GenBank mRNA | AF099974.1 | - | 2 | 547 | ||
| AK133208.1 | - | 2 | 1899 | |||
| BC132627.1 | - | 2 | 485 | 40130904 | ||
| BC138070.1 | - | 2 | 485 | 8861089 | ||
| BC145134.1 | - | 3 | 652 | 9053682 | ||

Slfn3
Genomic Map