siRNA Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145356.3 | NP_663331.1 | 5 | 1829 | ||
| GenBank mRNA | AK045522.1 | - | 5 | 1793 | ||
| AK156372.1 | - | 3 | 1498 | |||
| BC018187.1 | - | 3 | 1526 | 4208891 | ||
| BC070424.1 | - | 5 | 1829 | 6837447 | ||

Zbtb7c
Genomic Map