siRNA Details
| Gene Symbol: Zfp648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204908.1 | NP_001191837.1 | 2 | 1596 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp648| Gene Symbol: Zfp648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204908.1 | NP_001191837.1 | 2 | 1596 | ||