siRNA Details
Gene Symbol: Cyp4f17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101445.1 | NP_001094915.1 | 10 | 1308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp4f17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101445.1 | NP_001094915.1 | 10 | 1308 |