siRNA Details
| Gene Symbol: AU022751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166433.1 | NP_001159905.1 | 1 | 357 | ||
| GenBank mRNA | AK087702.1 | - | 1 | 357 | ||
| BC100377.1 | - | 1 | 343 | 30938251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AU022751| Gene Symbol: AU022751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166433.1 | NP_001159905.1 | 1 | 357 | ||
| GenBank mRNA | AK087702.1 | - | 1 | 357 | ||
| BC100377.1 | - | 1 | 343 | 30938251 | ||