siRNA Details
| Gene Symbol: Zfp710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145999.1 | NP_001139471.1 | 2 | 402 | ||
| NM_001146000.1 | NP_001139472.1 | 2 | 402 | |||
| NM_175433.5 | NP_780642.3 | 2 | 485 | |||
| GenBank mRNA | AK083979.1 | - | 2 | 327 | ||
| AK154871.1 | - | 2 | 383 | |||
| AK154953.1 | - | 2 | 403 | |||
| AK155416.1 | - | 2 | 464 | |||
| BC058640.1 | - | 2 | 296 | 6835601 | ||
| BC060641.1 | - | 2 | 379 | 6849964 | ||

Zfp710
Genomic Map