siRNA Details
| Gene Symbol: Sycp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011517.2 | NP_035647.2 | 2 | 191 | ||
| GenBank mRNA | AK131949.1 | - | 2 | 115 | ||
| BC048483.1 | - | 2 | 114 | |||
| BC089540.1 | - | 2 | 114 | |||
| BC099552.1 | - | 2 | 114 | |||
| BC132079.1 | - | 2 | 192 | 40130356 | ||
| BC138509.1 | - | 2 | 192 | 8861529 | ||
| Y08485.1 | - | 2 | 200 | |||

Sycp3
Genomic Map