siRNA Details
| Gene Symbol: Metrnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144797.3 | NP_659046.1 | 2 | 472 | ||
| GenBank mRNA | AK034855.1 | - | 2 | 473 | ||
| AK085880.1 | - | 2 | 473 | |||
| AK170800.1 | - | 2 | 473 | |||
| AK171200.1 | - | 2 | 218 | |||
| BC019776.1 | - | 2 | 427 | 3672224 | ||
| BC024445.1 | - | 2 | 487 | 4974030 | ||
| BC024497.1 | - | 475 | 4982563 | |||
| BC026646.1 | - | 2 | 466 | 4987595 | ||

Metrnl
Genomic Map