siRNA Details
| Gene Symbol: Ttbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162864.1 | NP_001156336.1 | 4 | 597 | ||
| GenBank mRNA | AK144536.1 | - | 3 | 331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttbk1| Gene Symbol: Ttbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162864.1 | NP_001156336.1 | 4 | 597 | ||
| GenBank mRNA | AK144536.1 | - | 3 | 331 | ||