siRNA Details
| Gene Symbol: Fbxw13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177598.3 | NP_808266.1 | 1 | 50 | ||
| GenBank mRNA | AK054339.1 | - | 1 | 51 | ||
| BC119542.1 | - | 1 | 37 | 40056515 | ||
| BC127237.1 | - | 1 | 37 | 40056507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw13| Gene Symbol: Fbxw13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177598.3 | NP_808266.1 | 1 | 50 | ||
| GenBank mRNA | AK054339.1 | - | 1 | 51 | ||
| BC119542.1 | - | 1 | 37 | 40056515 | ||
| BC127237.1 | - | 1 | 37 | 40056507 | ||