siRNA Details
| Gene Symbol: Dsg1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181680.1 | NP_859008.1 | 15 | 2376 | ||
| GenBank mRNA | AK034104.1 | - | 2319 | |||
| AY192159.1 | - | 15 | 2376 | |||
| AY314983.1 | - | 15 | 2276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsg1c| Gene Symbol: Dsg1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181680.1 | NP_859008.1 | 15 | 2376 | ||
| GenBank mRNA | AK034104.1 | - | 2319 | |||
| AY192159.1 | - | 15 | 2376 | |||
| AY314983.1 | - | 15 | 2276 | |||