siRNA Details
| Gene Symbol: Inpp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127363.1 | NP_001120835.1 | 3 | 479 | ||
| NM_183144.3 | NP_898967.2 | 3 | 479 | |||
| GenBank mRNA | AK083449.1 | - | 4 | 478 | ||
| AK157558.1 | - | 3 | 342 | |||
| AK171360.1 | - | 3 | 479 | |||
| AK171880.1 | - | 3 | 455 | |||
| BC056341.1 | - | 3 | 479 | 5703345 | ||

Inpp5a
Genomic Map