siRNA Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145382.4 | NP_663357.2 | 9, 10 | 3835 | ||
| GenBank mRNA | AK040568.1 | - | 3, 4 | 1963 | ||
| AK155237.1 | - | 9, 10 | 3829 | |||
| AK173303.1 | - | 9, 10 | 2684 | |||
| BC021381.1 | - | 3, 4 | 1425 | 5043282 | ||
| BC023015.1 | - | 3, 4 | 1367 | 5364758 | ||
| BC025483.1 | - | 9, 10 | 2694 | 5150988 | ||

Fam193b
Genomic Map