siRNA Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153783.4 | NP_722478.2 | 4 | 1106 | ||
| GenBank mRNA | AF226656.2 | - | 4 | 1125 | ||
| AK010469.1 | - | 3 | 756 | |||
| AK030664.1 | - | 4 | 1107 | |||
| AK159137.1 | - | 2 | 393 | |||
| AK178754.1 | - | 2 | 296 | |||
| AK186235.1 | - | 2 | 260 | |||
| AM049402.1 | - | 4 | 1098 | |||
| BC033913.1 | - | 3 | 370 | |||
| BC082783.1 | - | 4 | 1105 | 6475809 | ||

Paox
Genomic Map