siRNA Details
| Gene Symbol: Iffo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205173.1 | NP_001192102.1 | 4 | 1141 | ||
| NM_183148.3 | NP_898971.1 | 7 | 761 | |||
| GenBank mRNA | BC027170.1 | - | 3 | 287 | 4952810 | |
| BC100381.1 | - | 7 | 761 | |||
| BE860309.1 | - | 2 | 130 | |||
| CN718972.1 | - | 1 | 105 | |||
| EH105040.1 | - | 1 | 77 | |||

Iffo2
Genomic Map