siRNA Details
| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175440.4 | NP_780649.1 | 5 | 675 | ||
| GenBank mRNA | AF542056.1 | - | 5 | 611 | ||
| AK080281.1 | - | 5 | 676 | |||
| BC117009.1 | - | 5 | 658 | 40126328 | ||
| BC117011.1 | - | 5 | 658 | 40126330 | ||

Prss27
Genomic Map