siRNA Details
| Gene Symbol: Gm5941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034103.1 | NP_001029275.1 | 17 | |||
| GenBank mRNA | BC100438.1 | - | 17 | 30916603 | ||
| Gene Symbol: BC061195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC061195.1 | - | 30 | 6706856 | ||

Gm5941
Genomic Map