siRNA Details
| Gene Symbol: Tal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287388.1 | NP_001274317.1 | 5 | 834 | ||
| NM_011527.3 | NP_035657.1 | 5 | 817 | |||
| GenBank mRNA | AK144164.1 | - | 4 | 788 | ||
| AK157852.1 | - | 5 | 835 | |||
| BC063060.1 | - | 5 | 817 | 6826611 | ||
| M59764.1 | - | 4 | 765 | |||

Tal1
Genomic Map