siRNA Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291268.1 | NP_001278197.1 | 5 | 1177 | ||
| NM_001291269.1 | NP_001278198.1 | 5 | 886 | |||
| NM_009330.3 | NP_033356.2 | 5 | 1255 | |||
| GenBank mRNA | AB008174.1 | - | 5 | 1075 | ||
| AB008175.1 | - | 5 | 997 | |||
| AB008176.1 | - | 5 | 1075 | |||
| AB008177.1 | - | 5 | 997 | |||
| AB052659.1 | - | 5 | 1075 | |||
| AK004837.2 | - | 5 | 1256 | |||
| AK019258.1 | - | 5 | 809 | |||
| BC025189.1 | - | 5 | 886 | 5044299 | ||
| X55842.1 | - | 5 | 1075 | |||

Hnf1b
Genomic Map