siRNA Details
| Gene Symbol: Tcf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114140.1 | NP_001107612.1 | 2 | 3883 | ||
| NM_013836.3 | NP_038864.3 | 2 | 3883 | |||
| GenBank mRNA | AK122239.1 | - | 1 | 2912 | ||
| AK133162.1 | - | 1 | 693 | |||
| AY007594.1 | - | 2 | 3883 | |||
| BC042485.1 | - | 2 | 1262 | |||
| BC080855.1 | - | 1 | 1009 | |||
| BC138038.1 | - | 2 | 3856 | 8861057 | ||
| BC171946.1 | - | 2 | 3856 | |||

Tcf20
Genomic Map