siRNA Details
| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161722.1 | NP_001155194.1 | 5 | 866 | ||
| NM_001161723.1 | NP_001155195.1 | 5 | 860 | |||
| NM_011549.3 | NP_035679.3 | 4 | 807 | |||
| GenBank mRNA | AF079095.1 | - | 4 | 788 | ||
| AK078755.1 | - | 5 | 798 | |||
| AK146016.1 | - | 4 | 832 | |||
| AK154971.1 | - | 5 | 864 | |||
| BC064789.1 | - | 4 | 807 | 30429545 | ||

Tfeb
Genomic Map