siRNA Details
| Gene Symbol: Fam173a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285982.1 | NP_001272911.1 | 3 | 213 | ||
| NM_001285984.1 | NP_001272913.1 | 3 | 213 | |||
| NM_145410.4 | NP_663385.2 | 3 | 498 | |||
| GenBank mRNA | AK132031.1 | - | 3 | 372 | ||
| AK146214.1 | - | 3 | 479 | |||
| AK162337.1 | - | 3 | 474 | |||
| BC008155.1 | - | 3 | 222 | 3154632 | ||
| BC096050.1 | - | 3 | 393 | 5119802 | ||

Fam173a
Genomic Map