siRNA Details
| Gene Symbol: Taar5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009574.1 | NP_001009574.1 | 1 | 821 | ||
| GenBank mRNA | BC139188.1 | - | 1 | 866 | 8862210 | |
| BC139191.1 | - | 1 | 866 | 8862213 | ||
More Information
Set Membership: |
|

Taar5
Genomic Map