siRNA Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172134.2 | NP_742146.1 | 3 | 281 | ||
| GenBank mRNA | AK039194.1 | - | 3 | 263 | ||
| AK080846.1 | - | 3 | 257 | |||
| AK145470.1 | - | 3 | 245 | |||
| AK166078.1 | - | 3 | 264 | |||
| AK166464.1 | - | 3 | 246 | |||
| BC027745.1 | - | 3 | 318 | 5064695 | ||
| BC032171.1 | - | 3 | 252 | |||
| BI143426.1 | - | 3 | 310 | |||

Pdxk
Genomic Map