siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183426.1 | NP_906271.1 | 14 | 1671 | ||
| GenBank mRNA | AJ512611.1 | - | 15 | 1827 | ||
| AK089496.1 | - | 10 | 1590 | |||
| AK149806.1 | - | 14 | 1695 | |||
| AK155462.1 | - | 14 | 1669 | |||
| AK155674.1 | - | 14 | 1668 | |||
| AK170733.1 | - | 12 | 1803 | |||
| BC056369.1 | - | 14 | 1671 | 6831600 | ||
| BC064113.1 | - | 11 | 1428 | |||

Sbno2
Genomic Map