siRNA Details
| Gene Symbol: Abhd17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145421.2 | NP_663396.1 | 4 | 1044 | ||
| GenBank mRNA | AK170785.1 | - | 4 | 1030 | ||
| AK220316.1 | - | 2 | 4291 | |||
| BC005632.1 | - | 4 | 1019 | 3964094 | ||
| BC082997.1 | - | 4 | 1008 | 4160194 | ||

Abhd17a
Genomic Map