siRNA Details
| Gene Symbol: Tmem11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168507.1 | NP_001161979.1 | 3 | 644 | ||
| NM_173453.3 | NP_775655.1 | 2 | 356 | |||
| GenBank mRNA | AK077600.1 | - | 2 | 352 | ||
| BC048514.1 | - | 2 | 104 | 6594842 | ||
| CX211768.1 | - | 3 | 378 | |||

Tmem11
Genomic Map