siRNA Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146027.2 | NP_666139.1 | 2 | 182 | ||
| GenBank mRNA | AK075869.1 | - | 2 | 146 | ||
| BC010784.1 | - | 2 | 133 | |||
| BC021346.1 | - | 2 | 151 | 4163541 | ||
| CN691874.1 | - | 2 | 182 | |||

Scrn2
Genomic Map