siRNA Details
| Gene Symbol: Nags | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145829.2 | NP_665828.1 | 7 | 1586 | ||
| GenBank mRNA | AF462069.1 | - | 7 | 1542 | ||
| AJ489814.1 | - | 7 | 1512 | |||
| AK075765.1 | - | 6 | 1232 | |||
| BC036161.1 | - | 4 | 2237 | |||
| BC057990.1 | - | 7 | 1550 | 5100021 | ||

Nags
Genomic Map